6-Amino-1-cyclopropylpyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080888-500MG 080888-1G 080888-5G
102671-768EA 387.69 USD
102671-768 102671-770 102671-772
6-Amino-1-cyclopropylpyrimidine-2,4(1H,3H)-dione
6-Amino-1-cyclopropylpyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD07690492
CAS Number: 75914-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR