1-{2-Cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethanone
Supplier: Matrix Scientific
080887-500MG
080887-1G
080887-5G
102671-762EA
258.46
USD
102671-762
102671-764
102671-766
1-{2-Cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethanone
1-{2-Cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethanone
Hazard: Irritant
Formula:
C₁₅H₁₉N₅O |
MDL Number:
MFCD22374996 CAS Number: 1379821-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...