Ethyl-7-amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Supplier: Matrix Scientific

080885-500MG 080885-1G 080885-5G
102671-750EA 258.46 USD
102671-750 102671-752 102671-754
Ethyl-7-amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-7-amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅N₅O₂
MDL Number: MFCD22374994
CAS Number: 1379811-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR