1-(2-Cyclobutyl-7-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-6-yl)ethanone
Supplier: Matrix Scientific
080883-500MG
080883-1G
080883-5G
102671-738EA
387.69
USD
102671-738
102671-740
102671-742
1-(2-Cyclobutyl-7-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-6-yl)ethanone
1-(2-Cyclobutyl-7-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-6-yl)ethanone
Hazard: Irritant
Formula:
C₁₂H₁₄N₄O |
MDL Number:
MFCD22374992 CAS Number: 1379811-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...