7-Chloro-2-cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Supplier: Matrix Scientific
080880-500MG
080880-1G
080880-5G
102671-720EA
387.69
USD
102671-720
102671-722
102671-724
7-Chloro-2-cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
7-Chloro-2-cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₄ |
MDL Number:
MFCD22374989 CAS Number: 1379811-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...