2-Cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one

Supplier: Matrix Scientific

080879-500MG 080879-1G 080879-5G
102671-714EA 387.69 USD
102671-714 102671-716 102671-718
2-Cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one
2-Cyclobutyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD22374988
CAS Number: 1379811-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR