3-Cyclobutyl-1H-1,2,4-triazol-5-amine

Supplier: Matrix Scientific

080878-500MG 080878-1G 080878-5G
102671-708EA 387.69 USD
102671-708 102671-710 102671-712
3-Cyclobutyl-1H-1,2,4-triazol-5-amine
3-Cyclobutyl-1H-1,2,4-triazol-5-amine

Hazard: Irritant

Formula: C₆H₁₀N₄
MDL Number: MFCD13188121
CAS Number: 496057-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR