5-Benzyl-2-(cyclopropylamino)-6-methylpyrimidin-4(3H)-one
Supplier: Matrix Scientific
080874-500MG
080874-1G
102671-688EA
221.54
USD
102671-688
102671-690
5-Benzyl-2-(cyclopropylamino)-6-methylpyrimidin-4(3H)-one
5-Benzyl-2-(cyclopropylamino)-6-methylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₅H₁₇N₃O |
MDL Number:
MFCD22374986 CAS Number: 1379811-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...