2-(Cyclopropylamino)-5,6-dimethylpyrimidin-4(3H)-one
Supplier: Matrix Scientific
080873-500MG
080873-1G
102671-684EA
387.69
USD
102671-684
102671-686
2-(Cyclopropylamino)-5,6-dimethylpyrimidin-4(3H)-one
2-(Cyclopropylamino)-5,6-dimethylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₉H₁₃N₃O |
MDL Number:
MFCD20293195 CAS Number: 1379811-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...