N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)-1,3-benzothiazol-2-amine
Supplier: Matrix Scientific
080871-500MG
080871-1G
080871-5G
102671-672EA
258.46
USD
102671-672
102671-674
102671-676
N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)-1,3-benzothiazol-2-amine
N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)-1,3-benzothiazol-2-amine
Hazard: Irritant
Formula:
C₁₃H₁₅N₅S |
MDL Number:
MFCD22374985 CAS Number: 1379811-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...