N-Cyclopropyl-4,6-dimethylpyrimidin-2-amine
Supplier: Matrix Scientific
080868-500MG
080868-1G
080868-5G
102671-654EA
387.69
USD
102671-654
102671-656
102671-658
N-Cyclopropyl-4,6-dimethylpyrimidin-2-amine
N-Cyclopropyl-4,6-dimethylpyrimidin-2-amine
Hazard: Irritant
Formula:
C₉H₁₃N₃ |
MDL Number:
MFCD12188765 CAS Number: 1154246-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...