Ethyl-2-(cyclopropylamino)-4-hydroxypyrimidine-5-carboxylate

Supplier: Matrix Scientific

080867-500MG 080867-1G 080867-5G
102671-648EA 258.46 USD
102671-648 102671-650 102671-652
Ethyl-2-(cyclopropylamino)-4-hydroxypyrimidine-5-carboxylate
Ethyl-2-(cyclopropylamino)-4-hydroxypyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₃
MDL Number: MFCD22374982
CAS Number: 1343491-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR