2-(Cyclopropylamino)-6-methylpyrimidin-4(3H)-one
Supplier: Matrix Scientific
080865-500MG
080865-1G
080865-5G
102671-636EA
258.46
USD
102671-636
102671-638
102671-640
2-(Cyclopropylamino)-6-methylpyrimidin-4(3H)-one
2-(Cyclopropylamino)-6-methylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₈H₁₁N₃O |
MDL Number:
MFCD20293194 CAS Number: 5734-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...