1-[2-(Cyclopropylamino)-4-methylpyrimidin-5-yl]ethanone
Supplier: Matrix Scientific
080864-500MG
080864-1G
080864-5G
102671-630EA
387.69
USD
102671-630
102671-632
102671-634
1-[2-(Cyclopropylamino)-4-methylpyrimidin-5-yl]ethanone
1-[2-(Cyclopropylamino)-4-methylpyrimidin-5-yl]ethanone
Hazard: Irritant
Formula:
C₁₀H₁₃N₃O |
MDL Number:
MFCD15202253 CAS Number: 1211473-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...