Ethyl-2-(cyclopropylamino)-4-methylpyrimidine-5-carboxylate

Supplier: Matrix Scientific

080862-500MG 080862-1G 080862-5G
102671-618EA 258.46 USD
102671-618 102671-620 102671-622
Ethyl-2-(cyclopropylamino)-4-methylpyrimidine-5-carboxylate
Ethyl-2-(cyclopropylamino)-4-methylpyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD22374981
CAS Number: 1379811-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR