Dimethyl-1-cyclopropyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

Supplier: Matrix Scientific

080861-500MG 080861-1G 080861-5G
102671-612EA 258.46 USD
102671-612 102671-614 102671-616
Dimethyl-1-cyclopropyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Dimethyl-1-cyclopropyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD22374980
CAS Number: 1379811-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR