5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

080860-500MG 080860-1G 080860-5G
102671-606EA 387.69 USD
102671-606 102671-608 102671-610
5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazine-2-thiol
5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₆H₁₁N₃S
MDL Number: MFCD22374979
CAS Number: 218929-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR