N-Cyclopropyl-1H-benzimidazol-2-amine
Supplier: Matrix Scientific
080857-500MG
080857-1G
080857-5G
102671-590EA
516.92
USD
102671-590
102671-592
102671-594
N-Cyclopropyl-1H-benzimidazol-2-amine
N-Cyclopropyl-1H-benzimidazol-2-amine
Hazard: Irritant
Formula:
C₁₀H₁₁N₃ |
MDL Number:
MFCD09754817 CAS Number: 941266-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...