tert-Butyl-4-(4-acetylphenyl)piperazine-1-carboxylate

Supplier: Matrix Scientific

080855-500MG 080855-1G 080855-5G
102671-578EA 166.15 USD
102671-578 102671-580 102671-582
tert-Butyl-4-(4-acetylphenyl)piperazine-1-carboxylate
tert-Butyl-4-(4-acetylphenyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₃
MDL Number: MFCD20039715
CAS Number: 189763-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR