4-(4-Acetylphenyl)piperazine-1-carboximidamide sulfate

Supplier: Matrix Scientific

080854-500MG 080854-1G 080854-5G
102671-572EA 193.85 USD
102671-572 102671-574 102671-576
4-(4-Acetylphenyl)piperazine-1-carboximidamide sulfate
4-(4-Acetylphenyl)piperazine-1-carboximidamide sulfate

Hazard: Irritant

Formula: C₁₃H₂₀N₄O₅S
MDL Number: MFCD22422009
CAS Number: 1417566-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR