N-[4-(3-Thienyl)-1,3-thiazol-2-yl]guanidine

Supplier: Matrix Scientific

080853-500MG 080853-1G
102671-568EA 516.92 USD
102671-568 102671-570
N-[4-(3-Thienyl)-1,3-thiazol-2-yl]guanidine
N-[4-(3-Thienyl)-1,3-thiazol-2-yl]guanidine

Hazard: Irritant

Formula: C₈H₈N₄S₂
MDL Number: MFCD22374975
CAS Number: 760903-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR