N-[4-(2-Thienyl)-1,3-thiazol-2-yl]guanidine

Supplier: Matrix Scientific

080852-500MG 080852-1G 080852-5G
102671-562EA 387.69 USD
102671-562 102671-564 102671-566
N-[4-(2-Thienyl)-1,3-thiazol-2-yl]guanidine
N-[4-(2-Thienyl)-1,3-thiazol-2-yl]guanidine

Hazard: Irritant

Formula: C₈H₈N₄S₂
MDL Number: MFCD18453759
CAS Number: 735225-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR