N-1,3-Benzodioxol-5-ylguanidine sulfate

Supplier: Matrix Scientific

080848-500MG 080848-1G 080848-5G
102671-542EA 387.69 USD
102671-542 102671-544 102671-546
N-1,3-Benzodioxol-5-ylguanidine sulfate
N-1,3-Benzodioxol-5-ylguanidine sulfate

Hazard: Irritant

Formula: C₈H₁₁N₃O₆S
MDL Number: MFCD22422007
CAS Number: 1417566-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR