N-(1,4,5,6-Tetrahydrocyclopenta-[d]imidazol-2-yl)guanidine sulfate

Supplier: Matrix Scientific

080847-500MG 080847-1G
102671-538EA 516.92 USD
102671-538 102671-540
N-(1,4,5,6-Tetrahydrocyclopenta-[d]imidazol-2-yl)guanidine sulfate
N-(1,4,5,6-Tetrahydrocyclopenta-[d]imidazol-2-yl)guanidine sulfate

Hazard: Irritant

Formula: C₇H₁₃N₅O₄S
MDL Number: MFCD22422006
CAS Number: 1417566-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR