N-Cyclopentylimidodicarbonimidic diamide sulfate

Supplier: Matrix Scientific

080845-500MG 080845-1G 080845-5G
102671-528EA 258.46 USD
102671-528 102671-530 102671-532
N-Cyclopentylimidodicarbonimidic diamide sulfate
N-Cyclopentylimidodicarbonimidic diamide sulfate

Hazard: Irritant

Formula: C₇H₁₇N₅O₄S
MDL Number: MFCD22422005
CAS Number: 1417567-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR