N-[4-(4-Fluorophenyl)-1,3-thiazol-2-yl]guanidine
Supplier: Matrix Scientific
080842-500MG
080842-1G
080842-5G
102671-512EA
258.46
USD
102671-512
102671-514
102671-516
N-[4-(4-Fluorophenyl)-1,3-thiazol-2-yl]guanidine
N-[4-(4-Fluorophenyl)-1,3-thiazol-2-yl]guanidine
Hazard: Irritant
Formula:
C₁₀H₉FN₄S |
MDL Number:
MFCD06739729 CAS Number: 2438-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...