N-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]guanidine

Supplier: Matrix Scientific

080841-500MG 080841-1G 080841-5G
102671-506EA 258.46 USD
102671-506 102671-508 102671-510
N-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]guanidine
N-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]guanidine

Hazard: Irritant

Formula: C₁₀H₉ClN₄S
MDL Number: MFCD02102731
CAS Number: 7120-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR