4-(2-Hydroxyethyl)piperazine-1-carboximidamide sulfate (salt)
Supplier: Matrix Scientific
080838-500MG
080838-1G
102671-494EA
193.85
USD
102671-494
102671-496
4-(2-Hydroxyethyl)piperazine-1-carboximidamide sulfate (salt)
4-(2-Hydroxyethyl)piperazine-1-carboximidamide sulfate (salt)
Hazard: Irritant
Formula:
C₇H₁₈N₄O₅S |
MDL Number:
MFCD01748422 CAS Number: 1394042-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...