4-Pyrimidin-2-ylpiperazine-1-carboximidamide sulfate

Supplier: Matrix Scientific

080837-500MG 080837-1G
102671-490EA 193.85 USD
102671-490 102671-492
4-Pyrimidin-2-ylpiperazine-1-carboximidamide sulfate
4-Pyrimidin-2-ylpiperazine-1-carboximidamide sulfate

Hazard: Irritant

Formula: C₉H₁₆N₆O₄S
MDL Number: MFCD22422004
CAS Number: 1417569-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR