4-(2-Furoyl)piperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080835-500MG
080835-1G
080835-5G
102671-480EA
258.46
USD
102671-480
102671-482
102671-484
4-(2-Furoyl)piperazine-1-carboximidamide sulfate
4-(2-Furoyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₀H₁₆N₄O₆S |
MDL Number:
MFCD22422002 CAS Number: 1417567-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...