4-Formylpiperazine-1-carboximidamide sulfate

Supplier: Matrix Scientific

080834-500MG 080834-1G 080834-5G
102671-474EA 387.69 USD
102671-474 102671-476 102671-478
4-Formylpiperazine-1-carboximidamide sulfate
4-Formylpiperazine-1-carboximidamide sulfate

Hazard: Irritant

Formula: C₆H₁₄N₄O₅S
MDL Number: MFCD22422001
CAS Number: 1417567-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR