Ethyl-4-[amino(imino)methyl]piperazine-1-carboxylate sulfate
Supplier: Matrix Scientific
080833-500MG
080833-1G
102671-470EA
258.46
USD
102671-470
102671-472
Ethyl-4-[amino(imino)methyl]piperazine-1-carboxylate sulfate
Ethyl-4-[amino(imino)methyl]piperazine-1-carboxylate sulfate
Hazard: Irritant
Formula:
C₈H₁₈N₄O₆S |
MDL Number:
MFCD01703699 CAS Number: 63981-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...