4-(3-Methoxyphenyl)piperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080831-500MG
080831-1G
080831-5G
102671-458EA
193.85
USD
102671-458
102671-460
102671-462
4-(3-Methoxyphenyl)piperazine-1-carboximidamide sulfate
4-(3-Methoxyphenyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₂H₂₀N₄O₅S MW: 332.38 g/mol |
MDL Number:
MFCD22421999 CAS Number: 1417567-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...