4-Phenylpiperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080826-500MG
080826-1G
080826-5G
102671-428EA
193.85
USD
102671-428
102671-430
102671-432
4-Phenylpiperazine-1-carboximidamide sulfate
4-Phenylpiperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₁H₁₈N₄O₄S |
MDL Number:
MFCD22421996 CAS Number: 24360-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...