4-Ethylpiperazine-1-carboximidamide sulfate

Supplier: Matrix Scientific

080825-500MG 080825-1G 080825-5G
102671-422EA 169.23 USD
102671-422 102671-424 102671-426
4-Ethylpiperazine-1-carboximidamide sulfate
4-Ethylpiperazine-1-carboximidamide sulfate

Hazard: Irritant

Formula: C₇H₁₈N₄O₄S
MDL Number: MFCD22421995
CAS Number: 1266376-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR