4-(1,3-Benzodioxol-5-ylmethyl)piperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080823-500MG
080823-1G
080823-5G
102671-410EA
193.85
USD
102671-410
102671-412
102671-414
4-(1,3-Benzodioxol-5-ylmethyl)piperazine-1-carboximidamide sulfate
4-(1,3-Benzodioxol-5-ylmethyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₃H₂₀N₄O₆S |
MDL Number:
MFCD22421993 CAS Number: 1417568-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...