4-Benzylpiperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080822-500MG
080822-1G
080822-5G
102671-404EA
193.85
USD
102671-404
102671-406
102671-408
4-Benzylpiperazine-1-carboximidamide sulfate
4-Benzylpiperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₂H₂₀N₄O₄S |
MDL Number:
MFCD01748417 CAS Number: 53502-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...