N-(6-Ethoxy-1,3-benzothiazol-2-yl)guanidine

Supplier: Matrix Scientific

080819-500MG 080819-1G 080819-5G
102671-388EA 387.69 USD
102671-388 102671-390 102671-392
N-(6-Ethoxy-1,3-benzothiazol-2-yl)guanidine
N-(6-Ethoxy-1,3-benzothiazol-2-yl)guanidine

Hazard: Irritant

Formula: C₁₀H₁₂N₄OS
MDL Number: MFCD22374972
CAS Number: 1379811-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR