N-(6-Phenoxy-1,3-benzothiazol-2-yl)guanidine

Supplier: Matrix Scientific

080817-500MG 080817-1G 080817-5G
102671-376EA 258.46 USD
102671-376 102671-378 102671-380
N-(6-Phenoxy-1,3-benzothiazol-2-yl)guanidine
N-(6-Phenoxy-1,3-benzothiazol-2-yl)guanidine

Hazard: Irritant

Formula: C₁₄H₁₂N₄OS
MDL Number: MFCD22374970
CAS Number: 1379811-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR