N-(6-Fluoro-1,3-benzothiazol-2-yl)guanidine
Supplier: Matrix Scientific
080816-500MG
080816-1G
080816-5G
102671-370EA
387.69
USD
102671-370
102671-372
102671-374
N-(6-Fluoro-1,3-benzothiazol-2-yl)guanidine
N-(6-Fluoro-1,3-benzothiazol-2-yl)guanidine
Hazard: Irritant
Formula:
C₈H₇FN₄S |
MDL Number:
MFCD22374969 CAS Number: 1379811-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...