N-(6-Chloro-1,3-benzothiazol-2-yl)guanidine
Supplier: Matrix Scientific
080815-500MG
080815-1G
080815-5G
102671-364EA
387.69
USD
102671-364
102671-366
102671-368
N-(6-Chloro-1,3-benzothiazol-2-yl)guanidine
N-(6-Chloro-1,3-benzothiazol-2-yl)guanidine
Hazard: Irritant
Formula:
C₈H₇ClN₄S |
MDL Number:
MFCD22374968 CAS Number: 50739-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...