N-(5,6-Dimethyl-1,3-benzothiazol-2-yl)guanidine
Supplier: Matrix Scientific
080814-500MG
080814-1G
102671-360EA
387.69
USD
102671-360
102671-362
N-(5,6-Dimethyl-1,3-benzothiazol-2-yl)guanidine
N-(5,6-Dimethyl-1,3-benzothiazol-2-yl)guanidine
Hazard: Irritant
Formula:
C₁₀H₁₂N₄S |
MDL Number:
MFCD22374967 CAS Number: 1379811-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...