N-(6-Ethyl-1,3-benzothiazol-2-yl)guanidine

Supplier: Matrix Scientific

080813-500MG 080813-1G 080813-5G
102671-354EA 387.69 USD
102671-354 102671-356 102671-358
N-(6-Ethyl-1,3-benzothiazol-2-yl)guanidine
N-(6-Ethyl-1,3-benzothiazol-2-yl)guanidine

Hazard: Irritant

Formula: C₁₀H₁₂N₄S
MDL Number: MFCD22374966
CAS Number: 1379811-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR