3,4-Dihydroisoquinoline-2(1H)-carboximidamide sulfate

Supplier: Matrix Scientific

080811-500MG 080811-1G 080811-5G
102671-342EA 387.69 USD
102671-342 102671-344 102671-346
3,4-Dihydroisoquinoline-2(1H)-carboximidamide sulfate
3,4-Dihydroisoquinoline-2(1H)-carboximidamide sulfate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₄S
MDL Number: MFCD00941496
CAS Number: 2514-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR