N-[Amino(imino)methyl]-4-phenylpiperazine-1-carboximidamide

Supplier: Matrix Scientific

080805-500MG 080805-1G 080805-5G
102671-308EA 258.46 USD
102671-308 102671-310 102671-312
N-[Amino(imino)methyl]-4-phenylpiperazine-1-carboximidamide
N-[Amino(imino)methyl]-4-phenylpiperazine-1-carboximidamide

Hazard: Irritant

Formula: C₁₂H₁₈N₆
MDL Number: MFCD15203589
CAS Number: 786578-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR