N-[Amino(imino)methyl]-4-benzylpiperazine-1-carboximidamide

Supplier: Matrix Scientific

080804-500MG 080804-1G 080804-5G
102671-302EA 258.46 USD
102671-302 102671-304 102671-306
N-[Amino(imino)methyl]-4-benzylpiperazine-1-carboximidamide
N-[Amino(imino)methyl]-4-benzylpiperazine-1-carboximidamide

Hazard: Irritant

Formula: C₁₃H₂₀N₆
MDL Number: MFCD22374964
CAS Number: 746564-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR