N-[Amino(imino)methyl]-4-methylpiperazine-1-carboximidamide
Supplier: Matrix Scientific
080802-500MG
080802-1G
080802-5G
102671-290EA
167.69
USD
102671-290
102671-292
102671-294
N-[Amino(imino)methyl]-4-methylpiperazine-1-carboximidamide
N-[Amino(imino)methyl]-4-methylpiperazine-1-carboximidamide
Hazard: Irritant
Formula:
C₇H₁₆N₆ |
MDL Number:
MFCD15204029 CAS Number: 18413-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...