N-[Amino(imino)methyl]piperidine-1-carboximidamide

Supplier: Matrix Scientific

080801-500MG 080801-1G 080801-5G
102671-284EA 221.54 USD
102671-284 102671-286 102671-288
N-[Amino(imino)methyl]piperidine-1-carboximidamide
N-[Amino(imino)methyl]piperidine-1-carboximidamide

Hazard: Irritant

Formula: C₇H₁₅N₅
MDL Number: MFCD00460679
CAS Number: 4705-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR