N-[Amino(imino)methyl]-2-methylindoline-1-carboximidamide

Supplier: Matrix Scientific

080800-500MG 080800-1G 080800-5G
102671-278EA 258.46 USD
102671-278 102671-280 102671-282
N-[Amino(imino)methyl]-2-methylindoline-1-carboximidamide
N-[Amino(imino)methyl]-2-methylindoline-1-carboximidamide

Hazard: Irritant

Formula: C₁₁H₁₅N₅
MDL Number: MFCD02700384
CAS Number: 326010-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR