N-[Amino(imino)methyl]indoline-1-carboximidamide

Supplier: Matrix Scientific

080799-500MG 080799-1G 080799-5G
102671-272EA 258.46 USD
102671-272 102671-274 102671-276
N-[Amino(imino)methyl]indoline-1-carboximidamide
N-[Amino(imino)methyl]indoline-1-carboximidamide

Hazard: Irritant

Formula: C₁₀H₁₃N₅
MDL Number: MFCD00600606
CAS Number: 411211-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR